Learn More
Cole Parmer Oakton Hygrothermograph Chart Recorder Replacement Pens
Product Code. 15991227
Click to view available options
Quantity:
5/Pk.
Unit Size:
Pack of 5
Product Code. 15991227
Brand: Cole Parmer 4900653
Only null left
Add to basket
Add to basket
Different styles and colors to fit a wide variety of recorders and applications.
Specifications
Product Type Replacement Pen
Color Blue
Includes (1 m (3.3 ft.) cable with BNC connector
For Use With (Application) Hygrothermograph Charts
Quantity 5/Pk.