missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OAS2 Antibody (OTI6E6), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73137IR
This item is not returnable.
View return policy
Description
OAS2 Monoclonal specifically detects OAS2 in Human samples. It is validated for Western Blot.
Specifications
| OAS2 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| OAS2 | |
| Recombinant protein fragment corresponding to amino acids 60-335 of human OAS2 (NP_002526) produced in E.coli. | |
| 0.1 mL | |
| 4939.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI6E6 | |
| Western Blot | |
| (2-5′)oligo(A) synthase 2, (2′-5′)oligo(A) synthetase 2, (2-5′)oligo(A) synthetase 2, 2′5′ oligoadenylate synthetase 2, 2-5A synthase 2, 2-5A synthetase 2, 2′-5′-oligoadenylate synthase 2, 2′-5′-oligoadenylate synthetase 2 (69-71 kD), 2′-5′-oligoadenylate synthetase 2, 69/71kDa, EC 2.7.7, EC 2.7.7.-, MGC78578, p69 OAS/ p71 OAS, p69OAS/ p71OAS | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction