missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OAS2 Polyclonal Antibody

Product Code. 17783124
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17783124 20 μL 20µL
17740334 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17783124 Supplier Invitrogen™ Supplier No. PA5121730

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

OAS2 Polyclonal Antibody for Western Blot, IHC (P), ICC/IF

This gene encodes a member of the 2-5A synthetase family, essential proteins involved in the innate immune response to viral infection. The encoded protein is induced by interferons and uses adenosine triphosphate in 2'-specific nucleotidyl transfer reactions to synthesize 2',5'-oligoadenylates. These molecules activate latent RNase L, which results in viral RNA degradation and the inhibition of viral replication. The three known members of this gene family are located in a cluster on chromosome 12. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OAS2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene OAS2
Gene Accession No. E9Q9A9, P29728
Gene Alias (2-5')oligo(A) synthase 2; (2'-5')oligo(A) synthetase 2; 2',5'-oligoadenylate synthetase-like 11; 2,5-oligoadenylate synthetase-like 11; 2'-5' oligoadenylate synthetase 2; 2'-5' oligoadenylate synthetase-like 11; 2-5A synthase 2; 2'-5'-oligoadenylate synthase 2; 2'-5'-oligoadenylate synthetase 2; 2'-5'-oligoadenylate synthetase 2, 69/71kDa; A synthetase 2; MGC78578; Oas2; Oasl11; p69 OAS / p71 OAS; p69OAS / p71OAS
Gene Symbols OAS2
Host Species Rabbit
Immunogen Recombinant protein 2',5'-Oligoadenylate Synthetase 2. The antigen corresponds to amino acid range 11-346 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246728, 4939
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.