Learn More
Invitrogen™ OAS3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597217
Description
Immunogen sequence: AASCYDHPCF LRGMGDPVQS WKGPGLPRAG CSGLGHPIQL DPNQKTPENS KSLNAVYPRA GSKPPSCPAP GPTGAASIVP SVPGMALDLS QIPTKELDRF IQDHLKPSPQ FQEQVKKAID IILRCLHENC VHKASRVSKG GSFGRGTDLR DGCDVELIIF LNCFTDYKDQ GPRRAEILDE MRAQLESWWQ DQVPSLSLQF PEQNVPEALQ FQLVSTALKS WTDVSLLPAF DAVGQLSSGT KPNPQVYSRL LTSGCQEGEH KACFAELRRN FMNIRPVKLK NLILLVKHWY RQVAAQNKGK G; Positive Samples: U-87MG, DU145, HeLa; Cellular Location: Cytoplasm, Nucleus.
OAS3 is an interferon inducible protein that belongs to the 2-5A synthetase family, may play a role in mediating resistance to virus infection, control of cell growth, differentiation, and apoptosis. OAS3 synthesizes preferentially dimeric 2',5'-oligoadenylate molecules. GTP can be an alternative substrate. OAS3 binds double-stranded RNA and polymerizes ATP into PPP(A2'P5'A)N oligomers, which activate the latent RNase L that, when activated, cleaves single-stranded RNAs. The protein is present at high level in placenta trophoblast.
Specifications
| OAS3 | |
| Polyclonal | |
| Unconjugated | |
| OAS3 | |
| (2-5')oligo(A) synthase 3; (2-5')oligo(A) synthetase 3; 2',5'-oligoadenylate synthetase-like 10; 2'-5' oligoadenylate synthetase 3; 2'-5' oligoadenylate synthetase-like 10; 2-5A synthase 3; 2-5A synthetase 3; 2'-5'-oligoadenylate synthase 3; 2'-5'-oligoadenylate synthetase 3; 2'-5'-oligoadenylate synthetase 3, 100kDa; 2'-5'oligoadenylate synthetase p100; Oas3; oasl10; P/OKcl.4; p100; p100 OAS; p100OAS | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 246727, 4940, 494202 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.62 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q5MYT7, Q8VI93, Q9Y6K5 | |
| OAS3 | |
| Recombinant protein (or fragment) | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.