Learn More
Novus Biologicals™ OAT Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-29936
Product Code. 30048044
  • € 571.00 / 100µg
Description
OAT Polyclonal antibody specifically detects OAT in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OAT
Polyclonal
Tris-citrate/phosphate pH 7 -8.
Rabbit
Affinity purified
RUO
Primary
Human, Mouse
Purified
Immunoprecipitation, Western Blot
Unconjugated
GACR, HOGA, OAT ornithine aminotransferase, OATASE, OKT
Between 25 and 75
100 μg
metabolism, Proteases & Other Enzymes
4942
Store at 4°C. Do not freeze.