Learn More
OAT1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26574-1-AP-150UL
 View more versions of this product
Product Code. 16814995
  • € 410.00 / 150µL
Description
The OAT1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human OAT1. This antibody recognizes human, mouse antigen. The OAT1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
OAT1
Polyclonal
Unconjugated
150 μL
9356
IgG
Western Blot, Immunohistochemistry
333 μg/mL