missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OATP1 Polyclonal Antibody

Product Code. 15929824
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15929824 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15929824 Supplier Invitrogen™ Supplier No. PA567149

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: FLPNTLPKEGL ETNADIIKNE NEDKQKEEVK KEKYGITKDF LPFMKSLSCN Highest antigen sequence identity to the following orthologs - mouse 65%, rat 63%.

The organic anion transporting polypeptides, OATP-A (also designated OATP1, OATP1A2 and SLC21A3) and OATP-C (also designated OATP2, SLC21A6 and LST1), mediate hepatic uptake of cardiac glycosides. The expression of OATP-C, but not OATP-A, is inducible by phenobarbital and pregnenolone-16a-carbonitrile, resulting in the increased capacity of the liver to extract cardiac glycosides from the plasma. OATP-A is expressed in liver and kidney and helps mediate sodium-independent uptake of the anionic steroid conjugates dehydroepiandrosterone sulfate, estradiol-17-glucuronide and prostaglandin. OATP-C is exclusively expressed in liver and is localized to the basolateral hepatocyte membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OATP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLCO1A2
Gene Accession No. P46721
Gene Alias A530084B21; Brain digoxin carrier protein; brain-specific organic anion transporter; membrane transporter; OATP; Oatp1; OATP-1; Oatp1a1; OATP1A2; Oatp1a4; Oatp2; OATPA; OATP-A; OATP-B1; organic anion transporter 2; organic anion transporting polypeptide 1; organic anion transporting polypeptide 1a2; organic anion transporting polypeptide 1a2 variant 1; organic anion transporting polypeptide A; organic anion-transporting polypeptide 1; Slc21a1; SLC21A3; Slc21a5; Slco1a1; SLCO1A2; Slco1a4; SO1A2; Sodium-independent organic anion transporter; sodium-independent organic anion transporter 1; Sodium-independent organic anion-transporting polypeptide 1; sodium-independent organic anion-transporting polypeptide 2; solute carrier family (organic anion transporter) member 1; solute carrier family (organic anion transporter) member 3; solute carrier family 21 (organic anion transporter), member 1; solute carrier family 21 (organic anion transporter), member 3; solute carrier family 21 (organic anion transporter), member 5; solute carrier family 21 member 1; Solute carrier family 21 member 3; solute carrier family 21 member 5; solute carrier family 21, member 1; solute carrier organic anion transporter family member 1A1; Solute carrier organic anion transporter family member 1A2; solute carrier organic anion transporter family member 1A4; solute carrier organic anion transporter family, member 1a1; solute carrier organic anion transporter family, member 1A2; solute carrier organic anion transporter family, member 1a4
Gene Symbols SLCO1A2
Host Species Rabbit
Immunogen Recombinant Human OATP1. Recombinant protein control fragment (Product #RP-105520).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6579
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.