missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OATP1B3/SLCO1B3/OATP8 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98474F
This item is not returnable.
View return policy
Description
OATP1B3/SLCO1B3/OATP8 Polyclonal antibody specifically detects OATP1B3/SLCO1B3/OATP8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| OATP1B3/SLCO1B3/OATP8 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| FITC | |
| Liver-specific organic anion transporter 2, liver-specific organic anion transporter 3TM13, LST2, LST-2, LST3, OATP1B3LST-3TM13, OATP8, OATP-8, Organic anion transporter 8, organic anion transporter LST-3c, Organic anion-transporting polypeptide 8, SLC21A8, solute carrier family 21 (organic anion transporter), member 8, Solute carrier family 21 member 8, solute carrier organic anion transporter family member 1B3, solute carrier organic anion transporter family, member 1B3 | |
| Produced in rabbits immunized with E. coli-derived Human OATP1B3/SLCO1B3/OATP8 fragment. | |
| 0.1 mL | |
| Cancer | |
| 28234 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction