missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OBFC1 Monoclonal Antibody (OTI1C5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503274S
This item is not returnable.
View return policy
Description
OBFC1 Monoclonal antibody specifically detects OBFC1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| OBFC1 | |
| Monoclonal | |
| 0.35 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13200 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5708 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI1C5 | |
| Unconjugated | |
| PSMD2 | |
| 0610009H20Rik; 2310057J23Rik; AAF44; AAF-44; AI413458; alpha accessory factor 44; alpha-accessory factor of 44 kDa; bA541N10.2; CST complex subunit STN1; Obfc1; oligonucleotide/oligosaccharide binding fold containing 1; oligonucleotide/oligosaccharide-binding fold containing 1; oligonucleotide/oligosaccharide-binding fold-containing protein 1; replication protein A 32 kDa subunit; RPA-32; STN1; STN1 subunit of CST complex; STN1, CST complex subunit; suppressor of cdc thirteen homolog | |
| Full length human recombinant protein of human PSMD2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction