missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OBFC2A Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-26640B
This item is not returnable.
View return policy
Description
OBFC2A Polyclonal antibody specifically detects OBFC2A in Human samples. It is validated for Western Blot
Specifications
| OBFC2A | |
| Polyclonal | |
| Western Blot | |
| DKFZp667M1322, FLJ13624, FLJ22833, hSSB2, MGC111163, oligonucleotide/oligosaccharide-binding fold containing 2A, Oligonucleotide/oligosaccharide-binding fold-containing protein 2A, Sensor of single-strand DNA complex subunit B2, Sensor of ssDNA subunit B2, SOSS complex subunit B2, SOSS-B2, SSB2Single-stranded DNA-binding protein 2 | |
| The immunogen recognized by this antibody maps to a region between residue 150 to 200 of human oligonucleotide/oligosaccharide-binding fold containing 2A using the numbering given in entry NP_001026886.1 (GeneID 64859). | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 64859 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction