missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OBFC2B Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00079035-B01P
This item is not returnable.
View return policy
Description
OBFC2B Polyclonal antibody specifically detects OBFC2B in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| OBFC2B | |
| Polyclonal | |
| PBS (pH 7.4) | |
| LP3587, MGC2731, oligonucleotide/oligosaccharide-binding fold containing 2B, Oligonucleotide/oligosaccharide-binding fold-containing protein 2B, Sensor of single-strand DNA complex subunit B1, Sensor of ssDNA subunit B1, Single-stranded DNA-binding protein 1, SOSS complex subunit B1, SOSS-B1, SSB1hSSB1 | |
| OBFC2B (NP_076973, 1 a.a. - 211 a.a.) full-length human protein. MTTETFVKDIKPGLKNLNLIFIVLETGRVTKTKDGHEVRTCKVADKTGSINISVWDDVGNLIQPGDIIRLTKGYASVFKGCLTLYTGRGGDLQKIGEFCMVYSEVPNFSEPNPEYSTQQAPNKAVQNDSNPSASQPTTGPSAASPASENQNGNGLSAPPGPGGGPHPPHTPSHPPSTRITRSQPNHTPAGPPGPSSNPVSNGKETRRSSKR | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| NP_076973 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 79035 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction