missing translation for 'onlineSavingsMsg'
Learn More

OBFC2B Monoclonal Antibody (OTI1E11), TrueMAB™, OriGene

Product Code. p-200012663
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17550602 30 μL 30µL
17530602 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17550602 Supplier Origene Technologies Supplier No. TA504022S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

OBFC2B Monoclonal antibody specifically detects OBFC2B in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen OBFC2B
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1E11
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Nln
Gene Accession No. Q9BYT8
Gene Alias 2610036N15Rik; hSSB1; LP3587; Nabp2; nucleic acid binding protein 2; nucleic acid-binding protein 2; Obfc2b; oligonucleotide/oligosaccharide-binding fold containing 2B; oligonucleotide/oligosaccharide-binding fold-containing protein 2B; RGD1308158; Sensor of single-strand DNA complex subunit B1; sensor of ssDNA subunit B1; single strand DNA-binding protein 1; single-stranded DNA-binding protein 1; SOSS complex subunit B1; SOSS-B1; SSB1
Host Species Mouse
Immunogen Full length human recombinant protein of human NLN produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57486
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.