missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OBFC2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100654
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total OBFC2B.
Component of the SOSS complex, a multiprotein complex that functions downstream of the MRN complex to promote DNA repair and G2/M checkpoint. In the SOSS complex, acts as a sensor of single-stranded DNA that binds to single-stranded DNA, in particular to polypyrimidines. The SOSS complex associates with DNA lesions and influences diverse endpoints in the cellular DNA damage response including cell-cycle checkpoint activation, recombinational repair and maintenance of genomic stability. Required for efficient homologous recombination-dependent repair of double-strand breaks (DSBs) and ATM-dependent signaling pathways.
Specifications
| OBFC2B | |
| Polyclonal | |
| Unconjugated | |
| NABP2 | |
| 2610036N15Rik; hSSB1; LP3587; Nabp2; nucleic acid binding protein 2; nucleic acid-binding protein 2; Obfc2b; oligonucleotide/oligosaccharide-binding fold containing 2B; oligonucleotide/oligosaccharide-binding fold-containing protein 2B; RGD1308158; Sensor of single-strand DNA complex subunit B1; sensor of ssDNA subunit B1; single strand DNA-binding protein 1; single-stranded DNA-binding protein 1; SOSS complex subunit B1; SOSS-B1; SSB1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 362813, 69917, 79035 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q3SWT1, Q8R2Y9, Q9BQ15 | |
| NABP2 | |
| A synthesized peptide derived from human NABP2(Accession Q9BQ15), corresponding to amino acid residues P175-R211. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction