missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OBP2B Antibody (002), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90310CL1
This item is not returnable.
View return policy
Description
OBP2B Monoclonal antibody specifically detects OBP2B in Human samples. It is validated for ELISA
Specifications
| OBP2B | |
| Monoclonal | |
| CoraFluor 1 | |
| hOBPIIb, MGC119022, OBPIIb, odorant binding protein 2B, odorant-binding protein 2b, odorant-binding protein IIb, UNQ653/PRO1283 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human OBP2B (Accession#: NP_055396.1; Met1-His170). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 002 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 29989 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction