Learn More
Invitrogen™ OBP2B Recombinant Rabbit Monoclonal Antibody (002)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA530722
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human OBP2B.
OBP2B probably binds and transports small hydrophobic volatile molecules.
Specifications
| OBP2B | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9NPH6 | |
| OBP2B | |
| Recombinant Human OBP2B Protein (Met1-His170). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 002 | |
| Unconjugated | |
| OBP2B | |
| LCN14; OBP2B; OBPIIb; odorant binding protein 2B; odorant-binding protein 2b; Odorant-binding protein IIb; UNQ653/PRO1283 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 29989 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.