missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCAM/NCAM2 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97119AF647
This item is not returnable.
View return policy
Description
OCAM/NCAM2 Polyclonal specifically detects OCAM/NCAM2 in Human samples. It is validated for ELISA.
Specifications
| OCAM/NCAM2 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| Alexa Fluor 647 | |
| N-CAM-2, NCAM-2, NCAM21MGC51008, neural cell adhesion molecule 2, neural cell adhesion molecule 2 precursor10 | |
| Produced in rabbits immunized with purified, recombinant OCAM/NCAM2 (NP_004531.2; Met1-Asn697). | |
| 0.1 mL | |
| Growth and Development, Membrane Vesicle Markers, Neuronal Cell Markers | |
| 4685 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction