missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OCC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564212
This item is not returnable.
View return policy
Description
Immunogen sequence: MGCGNSTATS AGAGQGPAGA AKDVTEESVT EDDKRRNYGG VYVGLPSEAV NMVSSQTKTV RK Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 52%.
OCC-1 (Overexpressed in colon carcinoma-1) was initially identified as a novel human cDNA overexpressed in a colon carcinoma cell line. Elevated levels of OCC-1 mRNA were observed in three of eight colon carcinomas, suggesting that OCC-1 may be a hallmark of only a subset of colon carcinomas. It is highly expressed in skeletal muscle, kidney, placenta, and pancreas tissues, with low to no expression in heart, brain, lung and liver tissues. At least two different OCC-1 mRNA species are known to exist.
Specifications
| OCC1 | |
| Polyclonal | |
| Unconjugated | |
| C12orf75 | |
| adipogenesis down-regulated 3; AGD3; C12orf75; chromosome 12 open reading frame 75; OCC1; OCC-1; Overexpressed in colon carcinoma 1 protein; overexpressed in colon carcinoma-1; putative overexpressed in colon carcinoma-1 protein variant C | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 387882 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TAD7 | |
| C12orf75, OCC1 | |
| Recombinant protein corresponding to Human OCC1. Recombinant protein control fragment (Product #RP-103594). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction