Learn More
Occludin Antibody (8H8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26449-50ul
This item is not returnable.
View return policy
Description
Occludin Monoclonal antibody specifically detects Occludin in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry
Specifications
| Occludin | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry | |
| 8H8 | |
| Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
| BLCPMG, occludin | |
| A synthesized peptide derived from Human Occludin [UniProt Q16625] | |
| 50 μL | |
| Signal Transduction | |
| 100506658 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |