Learn More
Occludin Antibody (CL1608) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52973PCP
Product Code. 30294169
  • € 759.00 / 0.10mL
Description
Occludin Monoclonal antibody specifically detects Occludin in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Occludin
Monoclonal
PerCP
BLCPMG, occludin
This antibody was developed using a recombinant protein derived from Q16625, with the exact immunogen sequence remaining proprietary.
0.1 mL
Signal Transduction
100506658
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL1608
PBS
Mouse
Protein A purified
RUO
Primary
Human, Mouse