missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Occludin Antibody (rOCLN/8525), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20881C
This item is not returnable.
View return policy
Description
Occludin Monoclonal antibody specifically detects Occludin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Occludin | |
| Monoclonal | |
| DyLight 650 | |
| BLCPMG, occludin | |
| Recombinant human Occludin fragment around aa 282-415 (Exact sequence is proprietary) | |
| 0.1 mL | |
| Signal Transduction | |
| 100506658 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rOCLN/8525 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction