missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Occludin Antibody (rOCLN/8525), Novus Biologicals™
Mouse Monoclonal Antibody
€ 250.00 - € 494.00
Specifications
| Antigen | Occludin |
|---|---|
| Clone | rOCLN/8525 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18630894
|
Novus Biologicals
NBP3-20356-20ug |
20 μg |
€ 250.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18616914
|
Novus Biologicals
NBP3-20356-100ug |
100 μg |
€ 494.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Occludin Monoclonal antibody specifically detects Occludin in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Occludin | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Signal Transduction | |
| 10mM PBS with 0.05% BSA | |
| 100506658 | |
| IgG1 κ | |
| Protein A or G purified |
| rOCLN/8525 | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BLCPMG, occludin | |
| Recombinant human Occludin fragment around aa 282-415 (Exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title