Learn More
Oct-3/4 Antibody (240408), Alexa Fluor™ 405, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB1759V
Product Code. 30061765
  • € 652.00 / 0.10mL
Description
Oct-3/4 Monoclonal antibody specifically detects Oct-3/4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Oct-3/4
Monoclonal
Alexa Fluor 405
50mM Sodium Borate
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
5460
Store at 4C in the dark.
IgG2b
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
240408
Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
OCT3,Oct-3,OCT4,Oct-4,OCT4,Oct3/4,OTF3,OTF-3,OTF4,POU domain protein,POU domain, class 5, transcription factor 1,POU5F1
E. coli-derived recombinant human Oct-3/4, Met1-Asn265 (Met262Leu), Accession # Q01860
0.1 mL