missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oct-4A Antibody (03), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-12775H
This item is not returnable.
View return policy
Description
Oct-4A Monoclonal antibody specifically detects Oct-4A in Human samples. It is validated for Flow Cytometry
Specifications
| Oct-4A | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 5460 | |
| Store at 4C in the dark. | |
| IgG2a |
| Flow Cytometry | |
| 03 | |
| Flow Cytometry | |
| OCT3;Oct-3;OCT4;Oct-4;OCT4;Oct3/4;OTF3;OTF-3;OTF4;POU domain protein;POU domain, class 5, transcription factor 1;POU5F1 | |
| A synthetic peptide corresponding to the C-terminus of Human Oct-4A. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction