Learn More
Oct-4A Antibody (653108), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB17591S
Product Code. 30066416
  • € 652.00 / 0.10mL
Description
Oct-4A Monoclonal antibody specifically detects Oct-4A in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Oct-4A
Monoclonal
Alexa Fluor 750
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
5460
Store at 4C in the dark.
IgG2a
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
653108
Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
OCT3,Oct-3,OCT4,Oct-4,OCT4,Oct3/4,OTF3,OTF-3,OTF4,POU domain protein,POU domain, class 5, transcription factor 1,POU5F1
E. coli-derived recombinant human Oct-4A, Met1-Glu135, Accession # NP_002692
0.1 mL