missing translation for 'onlineSavingsMsg'
Learn More

Oct-7-enal, TRC

Product Code. 30331681
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30331681 500 mg 500mg
30348119 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30331681 Supplier Toronto Research Chemicals Supplier No. TRCB509005500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oct-7-enal
CAS 21573-31-9
Molecular Formula C8H14O
Quantity 500 mg
Synonym 7-Octen-1-al
InChI Formula InChI=1S/C8H14O/c1-2-3-4-5-6-7-8-9/h2,8H,1,3-7H2
SMILES C=CCCCCCC=O
IUPAC Name oct-7-enal
Molecular Weight (g/mol) 126.2
Formula Weight 126.1045
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.