missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCT2 Antibody (OCT2/2137), Novus Biologicals™
Mouse Monoclonal Antibody
€ 290.00 - € 504.00
Specifications
| Antigen | OCT2 |
|---|---|
| Clone | OCT2/2137 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18605747
|
Novus Biologicals
NBP2-75734-100ug |
100 μg |
€ 504.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18649546
|
Novus Biologicals
NBP2-75734-20ug |
20 μg |
€ 290.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OCT2 Monoclonal antibody specifically detects OCT2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein ArraySpecifications
| OCT2 | |
| 0.2 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| 10 mM PBS with 0.05% BSA | |
| homeobox protein, Lymphoid-restricted immunoglobulin octamer-binding protein NF-A2, OCT2Oct-2, Octamer-binding protein 2, Octamer-binding transcription factor 2, OTF-2, OTF2POU domain class 2, transcription factor 2, POU class 2 homeobox 2, POU domain, class 2, transcription factor 2 | |
| Recombinant fragment of human OCT2 protein (around aa 112-297) (exact sequence is proprietary) (Uniprot: P09086) | |
| Primary | |
| Store at 4°C. |
| OCT2/2137 | |
| Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P09086 | |
| 5452 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title