missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCT2 Antibody (OCT2/7073R) [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14146JF669
This item is not returnable.
View return policy
Description
OCT2 Monoclonal antibody specifically detects OCT2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| OCT2 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| homeobox protein, Lymphoid-restricted immunoglobulin octamer-binding protein NF-A2, OCT2Oct-2, Octamer-binding protein 2, Octamer-binding transcription factor 2, OTF-2, OTF2POU domain class 2, transcription factor 2, POU class 2 homeobox 2, POU domain, class 2, transcription factor 2 | |
| Recombinant fragment of human OCT2 protein (around aa 112-297) (exact sequence is proprietary) (Uniprot: P09086) | |
| 0.1 mL | |
| Cancer | |
| 5452 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| OCT2/7073R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction