missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCT2 Antibody (S07-8H6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20017-100ul
This item is not returnable.
View return policy
Description
OCT2 Monoclonal antibody specifically detects OCT2 in Human, Rat samples. It is validated for Western Blot
Specifications
| OCT2 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot | |
| S07-8H6 | |
| Unconjugated | |
| homeobox protein, Lymphoid-restricted immunoglobulin octamer-binding protein NF-A2, OCT2Oct-2, Octamer-binding protein 2, Octamer-binding transcription factor 2, OTF-2, OTF2POU domain class 2, transcription factor 2, POU class 2 homeobox 2, POU domain, class 2, transcription factor 2 | |
| A synthetic peptide of human OCT2 (Uniprot # P09086) | |
| 100 μg | |
| Cancer | |
| 5452 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction