missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCT4 Antibody (CL7621) [Alexa Fluor™ 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21158AF350
This item is not returnable.
View return policy
Description
2025-10-04 00:00:00 Monoclonal antibody specifically detects 2025-10-04 00:00:00 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| 2025-10-04 00:00:00 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| class 5, transcription factor 1, MGC22487, Oct-3, OCT3Oct4, Oct-4, Octamer-binding protein 3, otc-4, OTF3POU domain class 5, transcription factor 1, OTF4, POU class 5 homeobox 1, POU-type homeodomain-containing DNA-binding protein | |
| This antibody was developed using a synthetic peptide derived from Q01860, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cellular Markers, Chromatin Research, Glia Markers, Neuroscience, Oligodendrocyte Cell Markers, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators | |
| 5460 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| CL7621 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction