missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCT4 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2379DL594
This item is not returnable.
View return policy
Description
OCT4 Polyclonal antibody specifically detects OCT4 in Human, Mouse, Rat, Bovine, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| OCT4 | |
| Polyclonal | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| class 5, transcription factor 1, MGC22487, Oct-3, OCT3Oct4, Oct-4, Octamer-binding protein 3, otc-4, OTF3POU domain class 5, transcription factor 1, OTF4, POU class 5 homeobox 1, POU-type homeodomain-containing DNA-binding protein | |
| A synthetic peptide made to an internal portion of the human OCT4 protein (between residues 100-200) [UniProt# Q01860] | |
| 0.1 mL | |
| Cellular Markers, Chromatin Research, Glia Markers, Neuroscience, Oligodendrocyte Cell Markers, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators | |
| 5460 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction