missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
OCT4 Polyclonal antibody specifically detects OCT4 in Mouse samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Dilution | Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | class 5, transcription factor 1, MGC22487, Oct-3, OCT3Oct4, Oct-4, Octamer-binding protein 3, otc-4, OTF3POU domain class 5, transcription factor 1, OTF4, POU class 5 homeobox 1, POU-type homeodomain-containing DNA-binding protein |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse OCT4. |
| Form | Purified |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?