missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCT4 Antibody (OCT4/6847R), DyLight 680, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21058FR
This item is not returnable.
View return policy
Description
OCT4 Monoclonal antibody specifically detects OCT4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| OCT4 | |
| Monoclonal | |
| DyLight 680 | |
| class 5, transcription factor 1, MGC22487, Oct-3, OCT3Oct4, Oct-4, Octamer-binding protein 3, otc-4, OTF3POU domain class 5, transcription factor 1, OTF4, POU class 5 homeobox 1, POU-type homeodomain-containing DNA-binding protein | |
| Recombinant fragment of human OCT4 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cellular Markers, Chromatin Research, Glia Markers, Neuroscience, Oligodendrocyte Cell Markers, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators | |
| 5460 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| OCT4/6847R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction