Learn More
Macherey-Nagel™ Octadecyl Phase Polygosil™ C18
Product Code. 10595951 Shop All Macherey-Nagel Products
Click to view available options
Quantity:
10 g
100 g
Particle Size:
5 μm
7 μm
10 μm
Pore Size:
100 Å
1000 Å
300 Å
60 Å
Unit Size:
100g
10g
Product Code. 10595951
Brand: Macherey-Nagel™ 711560.10