missing translation for 'onlineSavingsMsg'
Learn More

Octadecylmagnesium chloride, 0.3M solution in THF, AcroSeal™, ACROS Organics™

Brand:  Acros Organics 436018000

Additional Details : CAS : 116980-66-6 Weight : 0.72000kg Transport : UN number : 2924 Chem class : 3 Pack group : II


Packaging AcroSeal™ Glass Bottle
Quantity 800mL

 View more versions of this product

Product Code. 11985971

  • € 264.00 / 800mL


Estimated Shipment: 09-09-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

116980-66-6
313.24
octadecylmagnesium chloride, magnesium,chlorooctadecyl, onjpxwzqvlkgiu-uhfffaoysa-m, octadecylmagnesium chloride solution, 0.5 m in thf, n-octadecylmagnesium chloride, 0.25m w/w in thf
magnesium;octadecane;chloride
C18H37ClMg
BNWJANSKVJZWMB-UHFFFAOYSA-M
4192511
CCCCCCCCCCCCCCCCC[CH2-].[Mg+2].[Cl-]

Specifications

Octadecylmagnesium chloride
−20°C
0.906
Brown to Yellow or Gray
116980-66-6,109-99-9
93.04
octadecylmagnesium chloride, magnesium,chlorooctadecyl, onjpxwzqvlkgiu-uhfffaoysa-m, octadecylmagnesium chloride solution, 0.5 m in thf, n-octadecylmagnesium chloride, 0.25m w/w in thf
BNWJANSKVJZWMB-UHFFFAOYSA-M
magnesium;octadecane;chloride
4192511
Solution, Any Turbidity Can Occur
0.9060g/mL
AcroSeal™ Glass Bottle
67.0°C
800mL
87.82
C18H37ClMg
Solubility in water: reacts.
CCCCCCCCCCCCCCCCC[CH2-].[Mg+2].[Cl-]
313.24
313.24
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Octadecylmagnesium chloride, 0.3M solution in THF

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS Documents
Certificates