Learn More
Octadecyltriethoxysilane, TRC
Specifications
Specifications
| Chemical Name or Material | Octadecyltriethoxysilane |
| CAS | 7399-00-0 |
| Molecular Formula | C24 H52 O3 Si |
| Quantity | 5 g |
| Synonym | Triethoxyoctadecylsilane, LS 6970, ODES, P 09775, S 12325, SIO 6645.0, Si 118, Stearyltriethoxysilane, n-Octadecyltriethoxysilane |
| InChI Formula | InChI=1S/C24H52O3Si/c1-5-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-28(25-6-2,26-7-3)27-8-4/h5-24H2,1-4H3 |
| SMILES | CCCCCCCCCCCCCCCCCC[Si](OCC)(OCC)OCC |
| IUPAC Name | triethoxy(octadecyl)silane |
| Molecular Weight (g/mol) | 416.753 |
| Formula Weight | 416.369 |