Learn More
Lenz Laborglasintrumente™ Octagonal Plate Stoppers
Octagonal stopper made from solid DURAN™ glass
Brand:  Lenz Laborglasintrumente™ 01400514
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11983341
  • € 86.44 / Pack of 10
Estimated Shipment: 03-06-2024
to see stock.
Add to basket
Description
Description
  • DIN 12252
  • Solid glass
  • Pack of 10
Specifications
Specifications
Solid
10
DURAN™
NS 14/23
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers