missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCTN1/SLC22A4 Antibody (892147), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10985L
This item is not returnable.
View return policy
Description
OCTN1/SLC22A4 Monoclonal antibody specifically detects OCTN1/SLC22A4 in Human samples. It is validated for Flow Cytometry
Specifications
| OCTN1/SLC22A4 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6583 | |
| Store at 4C in the dark. | |
| IgG2a |
| Flow Cytometry | |
| 892147 | |
| Flow Cytometry | |
| Ergothioneine transporter, ET transporter, ETT, MGC34546, MGC40524, OCTN1integral membrane transport protein, Organic cation/carnitine transporter 1, solute carrier family 22 (organic cation/ergothioneine transporter), member 4, solute carrier family 22 member 4, UT2H | |
| NS0 mouse myeloma cell line transfected with human SLC22A4, Accession # Q9H015 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction