missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCTN2/SLC22A5 Antibody (893753), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB11004MFV610
This item is not returnable.
View return policy
Description
OCTN2/SLC22A5 Monoclonal antibody specifically detects OCTN2/SLC22A5 in Human samples. It is validated for Flow Cytometry
Specifications
| OCTN2/SLC22A5 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 893753 | |
| Flow Cytometry | |
| CDSP, FLJ46769, High-affinity sodium-dependent carnitine cotransporter, OCTN2high-affinity sodium dependent carnitine cotransporter, OCTN2VT, organic cation transporter 2, organic cation transporter 5, Organic cation/carnitine transporter 2, SCD, solute carrier family 22 (organic cation transporter), member 5, solute carrier family 22 (organic cation/carnitine transporter), member 5, solute carrier family 22 member 5 | |
| NS0 transfected with human SLC22A5, Accession # O76082 | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 6584 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction