missing translation for 'onlineSavingsMsg'
Learn More

Octylonium Bromide, TRC

Product Code. 30420759
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30420759 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30420759 Supplier Toronto Research Chemicals Supplier No. TRCO293250500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Octylonium Bromide
CAS 26095-59-0
Purity Grade Notes HPLC
Molecular Formula C29 H43 N2 O4 . Br
Quantity 500 mg
Synonym Ethanaminium','N,N-diethyl-N-methyl-2-[[4-[[2-(octyloxy)benzoyl]amino]benzoyl]oxy]-','bromide (1:1), Ammonium','diethyl(2-hydroxyethyl)methyl-','bromide','p-[o-(octyloxy)benzamido]benzoate (8CI), Ethanaminium','N,N-diethyl-N-methyl-2-[[4-[[2-(octyloxy)benzoyl]amino]benzoyl]oxy]-','bromide (9CI), Benzoic acid','p-[o-(octyloxy)benzamido]-','ester with diethyl(2-hydroxyethyl)methylammonium bromide (8CI), Octylonium bromide, Otilonium bromide, SP 63, Spasmomen
InChI Formula InChI=1S/C29H42N2O4.BrH/c1-5-8-9-10-11-14-22-34-27-16-13-12-15-26(27)28(32)30-25-19-17-24(18-20-25)29(33)35-23-21-31(4,6-2)7-3;/h12-13,15-20H,5-11,14,21-23H2,1-4H3;1H
SMILES [Br-].CCCCCCCCOc1ccccc1C(=O)Nc2ccc(cc2)C(=O)OCC[N+](C)(CC)CC
IUPAC Name diethyl-methyl-[2-[4-[(2-octoxybenzoyl)amino]benzoyl]oxyethyl]azanium;bromide
Molecular Weight (g/mol) 563.57
Formula Weight 562.2406
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.