missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ODAM Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 450.00 - € 648.00
Specifications
| Antigen | ODAM |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18642045
|
Novus Biologicals
NBP3-21260-100ul |
100 μg |
€ 648.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18639654
|
Novus Biologicals
NBP3-21260-25ul |
25 μg |
€ 450.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ODAM Polyclonal antibody specifically detects ODAM in Human samples. It is validated for ImmunofluorescenceSpecifications
| ODAM | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| APIN, FLJ20513, odontogenic ameloblast-associated protein, odontogenic, ameloblast asssociated | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: IPQRLMSASNSNELLLNLNNGQLLPLQLQGPLNSWIPPFSGILQQQQQAQIPGLSQFSLSALDQFAGLLPNQIPLTGEASFAQGA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.2, 40% glycerol | |
| 54959 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title