missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ODC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102529
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ODC1.
Ornithine decarboxylase catalyzes the conversion of ornithine to putrescine in the first and apparently rate-limiting step in polyamine biosynthesis.
Specifications
| ODC1 | |
| Polyclonal | |
| Unconjugated | |
| Odc1 | |
| MGC103389; Odc; ODC1; Ornithine decarboxylase; ornithine decarboxylase 1; ornithine decarboxylase, structural 1; Ornitine decarboxylase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18263, 24609, 4953 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P00860, P09057, P11926 | |
| Odc1 | |
| A synthesized peptide derived from human ODC1(Accession P11926), corresponding to amino acid residues E7-K57. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction