missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleolin Antibody (364-5), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47861PCP
This item is not returnable.
View return policy
Description
Nucleolin Monoclonal antibody specifically detects Nucleolin in Human, Bovine (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Specifications
| Nucleolin | |
| Monoclonal | |
| PerCP | |
| C23, FLJ45706, FLJ59041, nucleolin, Protein C23 | |
| lysate of SU-DHL-1 Nuclei (Uniprot: P19338) | |
| 0.1 mL | |
| Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 4691 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 364-5 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Bovine (Negative), Mouse (Negative), Rat (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction