missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ODZ3 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81974H
This item is not returnable.
View return policy
Description
ODZ3 Polyclonal antibody specifically detects ODZ3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ODZ3 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| FLJ10474, FLJ10886, KIAA1455odd Oz/Ten-m homolog 3, odz, odd Oz/ten-m homolog 3 (Drosophila), ODZ3-like protein, Protein Odd Oz/ten-m homolog 3, ten-3, tenascin-M3, teneurin-3, Ten-m3, TNM3 | |
| ODZ3 antibody was raised against a 17 amino acid peptide near the amino terminus of human ODZ3. The immunogen is located within the first 50 amino acids of ODZ3. Amino Acid Squence: SLTKSRREKERRYTNS | |
| 0.1 mL | |
| Vision | |
| 55714 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction