missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OFD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570403
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 79%.
This gene is located on the X chromosome and encodes a centrosomal protein. A knockout mouse model has been used to study the effect of mutations in this gene. The mouse gene is also located on the X chromosome, however, unlike the human gene it is not subject to X inactivation. Mutations in this gene are associated with oral-facial-digital syndrome type I and Simpson-Golabi-Behmel syndrome type 2. Many pseudogenes have been identified; a single pseudogene is found on chromosome 5 while as many as fifteen have been found on the Y chromosome.
Specifications
| OFD1 | |
| Polyclonal | |
| Unconjugated | |
| OFD1 | |
| 71-7A; CXorf5; DXGgc7e; JBTS10; Ofd1; OFD1 centriole and centriolar satellite protein; ofd1 protein; OFD1, centriole and centriolar satellite protein; oral-facial-digital syndrome 1; oral-facial-digital syndrome 1 gene homolog; oral-facial-digital syndrome 1 gene homolog (human); oral-facial-digital syndrome 1 protein; Oral-facial-digital syndrome 1 protein homolog; ORF2; Protein 71-7A; putative oral-facial-digital syndrome 1 protein; RGD1562231; RP23; SGBS2; wu:fa07a10; wu:fd17e11; zgc:92562 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 302661 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| OFD1 | |
| synthetic peptide directed towards the middle region of human OFD1. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction