Learn More
OGDH Mouse anti-Human, Unconjugated, Clone: 1H7A2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66285-1-IG-20UL
This item is not returnable.
View return policy
Description
The OGDH antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human OGDH. This antibody recognizes human, rat antigen. The OGDH antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| OGDH | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 1H7A2 | |
| Unconjugated | |
| 20 μL | |
| 4967 | |
| IgG2b |