missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OGFOD1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-76532JF549
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
OGFOD1 Polyclonal antibody specifically detects OGFOD1 in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifica
| OGFOD1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA | |
| Janelia Fluor 549 | |
| 2-oxoglutarate and iron-dependent oxygenase domain containing 1,2-oxoglutarate and iron-dependent oxygenase domain-containing protein 1, EC 1.14.11, FLJ10826, KIAA1612TPA1, termination and polyadenylation 1, homolog, Termination and polyadenylation 1 homolog, TPA1EC 1.14.11.- | |
| Antibody was raised against a 18 amino acid synthetic peptide near the carboxy terminus of human OGFOD1. The immunogen is located within the first 50 amino acids of OGFOD1. Amino Acid Squence: ETLKKQVAEAWSRRTPFS | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 55239 | |
| Store at 4°C in the dark. | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto