missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OGFOD1 Antibody (OTI1D9), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73145APC
This item is not returnable.
View return policy
Description
OGFOD1 Monoclonal specifically detects OGFOD1 in Human, Monkey samples. It is validated for Flow Cytometry.
Specifications
| OGFOD1 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| OGFOD1 | |
| Full length recombinant protein of human OGFOD1 (NP_060703) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Monkey | |
| IgG1 |
| Flow Cytometry | |
| OTI1D9 | |
| Flow Cytometry | |
| 2-oxoglutarate and iron-dependent oxygenase domain containing 1,2-oxoglutarate and iron-dependent oxygenase domain-containing protein 1, EC 1.14.11, FLJ10826, KIAA1612TPA1, termination and polyadenylation 1, homolog, Termination and polyadenylation 1 homolog, TPA1EC 1.14.11.- | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 55239.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction