missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OGFOD1 Antibody (OTI1D9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-00528
This item is not returnable.
View return policy
Description
OGFOD1 Monoclonal specifically detects OGFOD1 in Human, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| OGFOD1 | |
| Monoclonal | |
| 0.41 mg/mL | |
| Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:150 | |
| OGFOD1 | |
| Full length human recombinant protein of human OGFOD1 (NP_060703) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Monkey | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI1D9 | |
| Unconjugated | |
| 2-oxoglutarate and iron-dependent oxygenase domain containing 1,2-oxoglutarate and iron-dependent oxygenase domain-containing protein 1, EC 1.14.11, FLJ10826, KIAA1612TPA1, termination and polyadenylation 1, homolog, Termination and polyadenylation 1 homolog, TPA1EC 1.14.11.- | |
| Mouse | |
| 63.1 kDa | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 55239 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction