missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OGFOD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113038
This item is not returnable.
View return policy
Description
OGFOD2 Polyclonal Antibody for Western Blot, ELISA
OGFOD2 is a member of the 2-OG-Fe(II) dioxygenase family which plays an important role in ischemic survival and stress. Like the related protein OGFOD1, OGFOD2 is an oxidoreductase containing a Prolyl 4-hydroxylase alpha subunit. Little is known about the function of OGFOD2, but its similarity to OGFOD1 suggests that it is a novel stress granule component that may play an important proapoptotic role in the regulation of translation and HRI-mediated phosphorylation of eIF2alpha in cells.
Specifications
| OGFOD2 | |
| Polyclonal | |
| Unconjugated | |
| OGFOD2 | |
| 1300006G11Rik; 2-oxoglutarate and iron dependent oxygenase domain containing 2; 2-oxoglutarate and iron-dependent oxygenase domain containing 2; 2-oxoglutarate and iron-dependent oxygenase domain-containing protein 2; 5730405M13Rik; AU041636; AW552395; OGFOD2; RGD1310594 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 66627, 79676 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q6N063, Q9CQ04 | |
| OGFOD2 | |
| Recombinant Human 2-oxoglutarate and iron-dependent oxygenase domain-containing protein 2 protein (1-186AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction