missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OGG1 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-106V
This item is not returnable.
View return policy
Description
OGG1 Polyclonal specifically detects OGG1 in Human, Mouse, Rat, Porcine, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Flow (Intracellular).
Specifications
| OGG1 | |
| Polyclonal | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Flow (Intracellular) | |
| O15527 | |
| OGG1 | |
| A peptide derived from human Ogg1 (within amino acids 1-100). [UniProt# O15527] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Primate, Rabbit | |
| IgG |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| 8-hydroxyguanine DNA glycosylase, 8-oxoguanine DNA glycosylase, AP lyase, DNA-apurinic or apyrimidinic site lyase, HOGG1, MMH, MUTMOGH1HMMH, N-glycosylase/DNA lyase | |
| Rabbit | |
| 39 kDa | |
| 0.1 mL | |
| Base Excision Repair, Cancer, Chromatin Research, DNA Repair | |
| 4968 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction